3-Chloro-1H-pyrazolo[3,4-c]pyridine

Lieferant: Apollo Scientific
Gefahr

OR305367-250MG
APOSOR305367-250MGEA 201 EUR
APOSOR305367-250MG
3-Chloro-1H-pyrazolo[3,4-c]pyridine
3-Chloro-1H-pyrazolo[3,4-c]pyridine
Formel: C₆H₄ClN₃
Molecular Weight: 153,569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11846323
CAS-Nummer: 76006-14-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR