CCT128930

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[(4-chlorophenyl)methyl]-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-amine, 4-(4-chlorobenzyl)-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidin-4-amine, CCT-128930, CCT 128930, CCT128930

OR305371-250MG
APOSOR305371-250MGEA 812 EUR
APOSOR305371-250MG
CCT128930
CCT128930
Formel: C₁₈H₂₀ClN₅
Molecular Weight: 341,84 g/mol
Schmelzpunkt: 214…216 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13177916
CAS-Nummer: 885499-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR