6,7-Dihydro-5H-pyrrolo[3,4-b]pyridin-5-one 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6,7-Dihydro-5-oxo-5H-pyrrolo[3,4-b]pyridine

OR305375-250MG
APOSOR305375-250MGEA 144 EUR
APOSOR305375-250MG
6,7-Dihydro-5H-pyrrolo[3,4-b]pyridin-5-one 95%
6,7-Dihydro-5H-pyrrolo[3,4-b]pyridin-5-one
Formel: C₇H₆N₂O
Molecular Weight: 134,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12923779
CAS-Nummer: 40107-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR