5-Pyrimidinecarbonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR305377-1G OR305377-5G
APOSOR305377-1GEA 0 EUR
APOSOR305377-1G APOSOR305377-5G
5-Pyrimidinecarbonyl chloride
5-Pyrimidinecarbonyl chloride
Formel: C₅H₃ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD03789627
CAS-Nummer: 40929-48-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR