Pyrimidine-5-carbohydrazide

Lieferant: Apollo Scientific

OR305378-250MG
APOSOR305378-250MGEA 335 EUR
APOSOR305378-250MG
Pyrimidine-5-carbohydrazide
Pyrimidine-5-carbohydrazide
Formel: C₅H₆N₄O
Molecular Weight: 138,127 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09951968
CAS-Nummer: 40929-42-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR