5-Iodopyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
OR305385-1G
APOSOR305385-1GEA
31.9
EUR
APOSOR305385-1G
5-Iodopyrimidine
5-Iodopyrimidine
Formel:
C4H3IN2 Schmelzpunkt: 126 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00233961 CAS-Nummer: 31462-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...