tert-Butyl 5-bromo-3-methyl-1H-pyrazolo[3,4-b]pyridine-1-carboxylate
Lieferant: Apollo Scientific
OR305390-250MG
APOSOR305390-250MGEA
142
EUR
APOSOR305390-250MG
tert-Butyl 5-bromo-3-methyl-1H-pyrazolo[3,4-b]pyridine-1-carboxylate
tert-Butyl 5-bromo-3-methyl-1H-pyrazolo[3,4-b]pyridine-1-carboxylate
Formel:
C₁₂H₁₄BrN₃O₂ Molecular Weight: 312,162 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13183735 CAS-Nummer: 916258-24-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...