1-(1H-Pyrazolo[3,4-b]pyridin-3-yl)ethanone

Lieferant: Apollo Scientific

OR305392-250MG
APOSOR305392-250MGEA 98.6 EUR
APOSOR305392-250MG
1-(1H-Pyrazolo[3,4-b]pyridin-3-yl)ethanone
1-(1H-Pyrazolo[3,4-b]pyridin-3-yl)ethanone
Formel: C₈H₇N₃O
Molecular Weight: 161,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13178075
CAS-Nummer: 889451-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR