4-Bromo-2-methoxypyrimidine

Lieferant: Apollo Scientific
Achtung

OR30540-250MG
APOSOR30540-250MGEA 29 EUR
APOSOR30540-250MG
4-Bromo-2-methoxypyrimidine
4-Bromo-2-methoxypyrimidine
Formel: C₅H₅BrN₂O
Molecular Weight: 189,0100 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09879097
CAS-Nummer: 959240-72-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR