(6-Aminopyridin-3-yl)acetic acid 95%

Lieferant: Apollo Scientific
Achtung

OR305409-250MG
APOSOR305409-250MGEA 0 EUR
APOSOR305409-250MG
(6-Aminopyridin-3-yl)acetic acid 95%
(6-Aminopyridin-3-yl)acetic acid
Formel: C₇H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09757480
CAS-Nummer: 39658-45-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR