Ethyl (3-aminopyridin-2-yl)acetate 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR305412-250MG
APOSOR305412-250MGEA
201
EUR
APOSOR305412-250MG
Ethyl (3-aminopyridin-2-yl)acetate 95%
Ethyl (3-aminopyridin-2-yl)acetate
Formel:
C₉H₁₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11616148 CAS-Nummer: 295327-27-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...