3-(Aminomethyl)-6-methyl-4-propylpyridin-2(1H)-one acetate

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Aminomethyl)-6-methyl-4-propylpyridin-2(1H)-one

OR305421-1G
APOSOR305421-1GEA 840 EUR
APOSOR305421-1G
3-(Aminomethyl)-6-methyl-4-propylpyridin-2(1H)-one acetate
3-(Aminomethyl)-6-methyl-4-propylpyridin-2(1H)-one acetate
Formel: C₁₀H₁₆N₂O·CH₃CO₂H
Molecular Weight: 240,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25541770
CAS-Nummer: 1346575-64-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR