Nicotinaldehyde N-oxide

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Formylpyridinium-1-olate, 3-Formylpyridine N-oxide, Pyridine-3-carboxaldehyde N-oxide

OR305425-250MG
APOSOR305425-250MGEA 173 EUR
APOSOR305425-250MG
Nicotinaldehyde N-oxide
Nicotinaldehyde N-oxide
Formel: C₆H₅NO₂
Molecular Weight: 123.109 g/mol
Schmelzpunkt: 130…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD13175267
CAS-Nummer: 22346-73-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR