[1,3]Thiazolo[4,5-b]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: Thiazolo[4,5-b]pyridine

OR305430-1G
APOSOR305430-1GEA 441 EUR
APOSOR305430-1G
[1,3]Thiazolo[4,5-b]pyridine
[1,3]Thiazolo[4,5-b]pyridine
Formel: C₆H₄N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD11878037
CAS-Nummer: 273-98-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR