[1,3]Thiazolo[5,4-b]pyridine-5-carboxylic acid
Lieferant: Apollo Scientific
OR305434-250MG
APOSOR305434-250MGEA
464
EUR
APOSOR305434-250MG
[1,3]Thiazolo[5,4-b]pyridine-5-carboxylic acid
[1,3]Thiazolo[5,4-b]pyridine-5-carboxylic acid
Formel:
C₇H₄N₂O₂S Molecular Weight: 180,184 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22555424 CAS-Nummer: 1304788-06-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...