2-Chloropyrimidine-4-carboxamide

Lieferant: Apollo Scientific
Achtung

OR30544-500MG
APOSOR30544-500MGEA 24.8 EUR
APOSOR30544-500MG
2-Chloropyrimidine-4-carboxamide
2-Chloropyrimidine-4-carboxamide
Formel: C₅H₄ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00234053
CAS-Nummer: 22536-66-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR