Methyl 6-chloro-1H-indazole-4-carboxylate

Lieferant: Apollo Scientific

OR305441-250MG
APOSOR305441-250MGEA 159 EUR
APOSOR305441-250MG
Methyl 6-chloro-1H-indazole-4-carboxylate
Methyl 6-chloro-1H-indazole-4-carboxylate
Formel: C₉H₇ClN₂O₂
Molecular Weight: 210,617 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781390
CAS-Nummer: 885519-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR