4-Chloro-2-methoxypyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
OR30545-100MG
APOSOR30545-100MGEA
24.8
EUR
APOSOR30545-100MG
4-Chloro-2-methoxypyrimidine
4-Chloro-2-methoxypyrimidine
Formel:
C₅H₅ClN₂O Molecular Weight: 144,5590 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09025734 CAS-Nummer: 51421-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...