1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid ethyl ester 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 1H-pyrazolo[4,3-b]pyridine-3-carboxylate

OR305461-250MG
APOSOR305461-250MGEA 150 EUR
APOSOR305461-250MG
1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid ethyl ester 97%
1H-Pyrazolo[4,3-b]pyridine-3-carboxylic acid ethyl ester
Formel: C₉H₉N₃O₂
Molecular Weight: 191,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16658534
CAS-Nummer: 1234616-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR