2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl

Lieferant: Apollo Scientific

OR305465-1G
APOSOR305465-1GEA 0 EUR
APOSOR305465-1G
2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl
2'-Bromo-5'-phenyl-1,1':3',1''-terphenyl
Formel: C₂₄H₁₇Br
Lagertemperatur: Raumtemperatur
MDL: MFCD00092320
CAS-Nummer: 10368-73-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR