8-Chloroimidazo[1,5-a]pyrazine 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR305494-250MG
OR305494-100MG
OR305494-1G
APOSOR305494-250MGEA
173
EUR
APOSOR305494-250MG
APOSOR305494-100MG
APOSOR305494-1G
8-Chloroimidazo[1,5-a]pyrazine 95%
8-Chloroimidazo[1,5-a]pyrazine
Formel:
C₆H₄ClN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20040069 CAS-Nummer: 56468-23-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...