3-Oxoindane-5-carboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 2,3-Dihydro-3-oxo-1H-indene-5-carboxylic acid, 3-Oxo-2,3-dihydro-1H-indene-5-carboxylic acid

OR30552-250MG
APOSOR30552-250MGEA 125 EUR
APOSOR30552-250MG
3-Oxoindane-5-carboxylic acid 97%
3-Oxoindane-5-carboxylic acid
Formel: C₁₀H₈O₃
Molecular Weight: 176,169 g/mol
Schmelzpunkt: 256…260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179295
CAS-Nummer: 60031-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR