5-Chloro-1H-pyrazolo[4,3-d]pyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
OR305533-250MG
APOSOR305533-250MGEA
27.7
EUR
APOSOR305533-250MG
5-Chloro-1H-pyrazolo[4,3-d]pyrimidine
5-Chloro-1H-pyrazolo[4,3-d]pyrimidine
Formel:
C₅H₃ClN₄ Molecular Weight: 154,557 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18251003 CAS-Nummer: 633328-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...