5,7-Dichloro-1H-pyrrolo[2,3-c]pyridine
Lieferant: Apollo Scientific
OR305545-250MG
APOSOR305545-250MGEA
137
EUR
APOSOR305545-250MG
5,7-Dichloro-1H-pyrrolo[2,3-c]pyridine
5,7-Dichloro-1H-pyrrolo[2,3-c]pyridine
Formel:
C₇H₄Cl₂N₂ Molecular Weight: 187,026 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11845567 CAS-Nummer: 1001412-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...