4-(5-{4-[(Pent-1-yl)oxy]phenyl}isoxazol-3-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(5-(4-(Pentyloxy)phenyl)isoxazol-3-yl)benzoic acid, 4-{5-[4-(Pentyloxy)phenyl]-1,2-oxazol-3-yl}benzoic acid, 3-(4-Carboxyphenyl)-5-[4-(pentyloxy)phenyl]-1,2-oxazole

OR305548-250MG
APOSOR305548-250MGEA 24.8 EUR
APOSOR305548-250MG
4-(5-{4-[(Pent-1-yl)oxy]phenyl}isoxazol-3-yl)benzoic acid
4-(5-{4-[(Pent-1-yl)oxy]phenyl}isoxazol-3-yl)benzoic acid
Formel: C₂₁H₂₁NO₄
Molecular Weight: 351,396 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20267356
CAS-Nummer: 179162-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR