4-Chloro-2-methyl[1,3]oxazolo[5,4-c]pyridine

Lieferant: Apollo Scientific
Achtung

OR305553-250MG
APOSOR305553-250MGEA 1210 EUR
APOSOR305553-250MG
4-Chloro-2-methyl[1,3]oxazolo[5,4-c]pyridine
4-Chloro-2-methyl[1,3]oxazolo[5,4-c]pyridine
Formel: C₇H₅ClN₂O
Molecular Weight: 168,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24848274
CAS-Nummer: 1354831-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR