4-Amino-2-methoxypyridin
Lieferant: Apollo Scientific
Synonyme:
2-Methoxypyridin-4-amine
OR30557-5G
APOSOR30557-5GEA
21.9
EUR
APOSOR30557-5G
4-Amino-2-methoxypyridin
4-Amino-2-methoxypyridine
Formel:
C₆H₈N₂O |
MDL:
MFCD06738657 CAS-Nummer: 20265-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...