3-Iodo-1,5-naphthyridine

Lieferant: Apollo Scientific

OR305571-250MG
APOSOR305571-250MGEA 242 EUR
APOSOR305571-250MG
3-Iodo-1,5-naphthyridine
3-Iodo-1,5-naphthyridine
Formel: C₈H₅IN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16628231
CAS-Nummer: 1228666-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR