3-Iodo-1H-pyrazolo[4,3-b]pyridine

Lieferant: Apollo Scientific

OR305572-1G
APOSOR305572-1GEA 259 EUR
APOSOR305572-1G
3-Iodo-1H-pyrazolo[4,3-b]pyridine
3-Iodo-1H-pyrazolo[4,3-b]pyridine
Formel: C₆H₄IN₃
Molecular Weight: 245,021 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10696864
CAS-Nummer: 633328-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR