3-Formyl-1H-indazole-5-carboxylic acid

Lieferant: Apollo Scientific

OR305574-1G
APOSOR305574-1GEA 177 EUR
APOSOR305574-1G
3-Formyl-1H-indazole-5-carboxylic acid
3-Formyl-1H-indazole-5-carboxylic acid
Formel: C₉H₆N₂O₃
Molecular Weight: 190,156 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656900
CAS-Nummer: 885519-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR