4-Brom-m-toluylsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Brom-3-methylbenzoesäure

OR3056-250MG OR3056-1G
APOSOR3056-250MGEA 21.9 EUR
APOSOR3056-250MG APOSOR3056-1G
4-Brom-m-toluylsäure
4-Brom-m-toluylsäure
Formel: C₈H₇BrO₂
Schmelzpunkt: 212 - 216 °C
MDL: MFCD00039526
CAS-Nummer: 7697-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR