4-Brom-m-toluylsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Brom-3-methylbenzoesäure
OR3056-250MG
OR3056-1G
APOSOR3056-250MGEA
21.9
EUR
APOSOR3056-250MG
APOSOR3056-1G
4-Brom-m-toluylsäure
4-Brom-m-toluylsäure
Formel:
C₈H₇BrO₂ Schmelzpunkt: 212 - 216 °C |
MDL:
MFCD00039526 CAS-Nummer: 7697-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...