1-Methyl-2-nitro-1H-imidazole-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR305606-1G
APOSOR305606-1GEA 659 EUR
APOSOR305606-1G
1-Methyl-2-nitro-1H-imidazole-5-carboxylic acid
1-Methyl-2-nitro-1H-imidazole-5-carboxylic acid
Formel: C₅H₅N₃O₄
Molecular Weight: 171,111 g/mol
Schmelzpunkt: 161…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD19443277
CAS-Nummer: 50700-55-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR