1-Benzothiophene-6-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Benzo[b]thiophene-6-carbaldehyde
OR305609-250MG
APOSOR305609-250MGEA
202
EUR
APOSOR305609-250MG
1-Benzothiophene-6-carboxaldehyde
1-Benzothiophene-6-carboxaldehyde
Formel:
C₉H₆OS Molecular Weight: 162,208 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18451824 CAS-Nummer: 6386-80-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...