1-(1-Benzothiophen-4-yl)piperazine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(Benzo[b]thiophen-4-yl)piperazine hydrochloride

OR305616-25G
APOSOR305616-25GEA 149 EUR
APOSOR305616-25G
1-(1-Benzothiophen-4-yl)piperazine hydrochloride
1-(1-Benzothiophen-4-yl)piperazine hydrochloride
Formel: C₁₂H₁₄N₂S·ClH
Molecular Weight: 254.779 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26743820
CAS-Nummer: 913614-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR