1,3-Benzothiazole-5-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Achtung

OR305622-250MG
APOSOR305622-250MGEA 690 EUR
APOSOR305622-250MG
1,3-Benzothiazole-5-sulphonyl chloride 95%
1,3-Benzothiazole-5-sulphonyl chloride
Formel: C₇H₄ClNO₂S₂
Molecular Weight: 233,695 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20233448
CAS-Nummer: 227278-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR