1,1,1-Triphenylsilanamine

Lieferant: Apollo Scientific
Achtung

OR305626-5G
APOSOR305626-5GEA 429 EUR
APOSOR305626-5G
1,1,1-Triphenylsilanamine
1,1,1-Triphenylsilanamine
Formel: C₁₈H₁₇NSi
Molecular Weight: 275,42 g/mol
Schmelzpunkt: 60…62 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00042816
CAS-Nummer: 4215-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR