1H-Pyrrolo[3,2-b]pyridine-5-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR305629-1G
OR305629-250MG
APOSOR305629-1GEA
0
EUR
APOSOR305629-1G
APOSOR305629-250MG
1H-Pyrrolo[3,2-b]pyridine-5-carbonitrile
1H-Pyrrolo[3,2-b]pyridine-5-carbonitrile
Formel:
C₈H₅N₃ Schmelzpunkt: 214…216 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09909831 CAS-Nummer: 146767-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...