Methyl bromo(2-cyanophenyl)acetate

Lieferant: Apollo Scientific
Gefahr

OR30563-100MG OR30563-1G OR30563-250MG
APOSOR30563-100MGEA 140 EUR
APOSOR30563-100MG APOSOR30563-1G APOSOR30563-250MG
Methyl bromo(2-cyanophenyl)acetate
Methyl bromo(2-cyanophenyl)acetate
Formel: C₁₀H₈BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04038664
CAS-Nummer: 885953-43-5
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR