5-Sulphanyl-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
OR305638-250MG
APOSOR305638-250MGEA
835
EUR
APOSOR305638-250MG
5-Sulphanyl-1H-indazole
5-Sulphanyl-1H-indazole
Formel:
C₇H₆N₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD13176737 CAS-Nummer: 569339-86-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...