8-(Hydroxymethyl)isoquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: (Isoquinolin-8-yl)methanol, 8-(Hydroxymethyl)-2-azanaphthalene

OR30566-500MG
APOSOR30566-500MGEA 284 EUR
APOSOR30566-500MG
8-(Hydroxymethyl)isoquinoline
8-(Hydroxymethyl)isoquinoline
Formel: C₁₀H₉NO
Molecular Weight: 159.185 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11869736
CAS-Nummer: 1159511-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR