8-(Bromomethyl)isoquinoline hydrobromide

Lieferant: Apollo Scientific
Achtung

OR30573-1G
APOSOR30573-1GEA 0 EUR
APOSOR30573-1G
8-(Bromomethyl)isoquinoline hydrobromide
8-(Bromomethyl)isoquinoline hydrobromide
Formel: C₁₀H₈BrN·BrH
Lagertemperatur: Raumtemperatur
MDL: MFCD12674806
CAS-Nummer: 1215541-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR