3-Bromothieno[3,2-c]pyridine

Lieferant: Apollo Scientific
Achtung

OR30582-250MG
APOSOR30582-250MGEA 79.9 EUR
APOSOR30582-250MG
3-Bromothieno[3,2-c]pyridine
3-Bromothieno[3,2-c]pyridine
Formel: C7H4BrNS
Molecular Weight: 214,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11869740
CAS-Nummer: 28783-18-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR