1-Methyl-1H-indazol-5-boronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Methyl-1H-indazol-5-yl-5-boronic acid, (1-methyl-1H-indazol-5-yl)boronic acid

OR30587-1G OR30587-250MG
APOSOR30587-1GEA 26.1 EUR
APOSOR30587-1G APOSOR30587-250MG
1-Methyl-1H-indazol-5-boronsäure
1-Methyl-1H-indazole-5-boronic acid
Formel: C₈H₉BN₂O₂
Molecular Weight: 175.9800 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09870052
CAS-Nummer: 590418-08-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR