2-Methyl-2H-indazol-5-boronsäurepinakolester

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-indazole

OR30597-250MG
APOSOR30597-250MGEA 39.3 EUR
APOSOR30597-250MG
2-Methyl-2H-indazol-5-boronsäurepinakolester
2-Methyl-2H-indazol-5-boronsäurepinakolester
Formel: C₁₄H₁₉BN₂O₂
Molecular Weight: 258,1239 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11109436
CAS-Nummer: 1189746-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR