8-Bromimidazo[1,2-a]pyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR30604-250MG
APOSOR30604-250MGEA
21.9
EUR
APOSOR30604-250MG
8-Bromimidazo[1,2-a]pyridin
8-Bromimidazo[1,2-a]pyridin
Formel:
C₇H₅BrN₂ Schmelzpunkt: 72…74 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08706060 CAS-Nummer: 850349-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...