4-(Oxetan-3-yloxy)benzoic acid
Lieferant: Apollo Scientific
OR306064-1G
APOSOR306064-1GEA
0
EUR
APOSOR306064-1G
4-(Oxetan-3-yloxy)benzoic acid
4-(Oxetan-3-yloxy)benzoic acid
Formel:
C₁₀H₁₀O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20233109 CAS-Nummer: 1349717-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...