1-(2-Methoxyethyl)-3-methyl-1H-pyrazol-5-amine 95%

Lieferant: Apollo Scientific
Achtung

OR306164-1G
APOSOR306164-1GEA 799 EUR
APOSOR306164-1G
1-(2-Methoxyethyl)-3-methyl-1H-pyrazol-5-amine 95%
1-(2-Methoxyethyl)-3-methyl-1H-pyrazol-5-amine
Formel: C₇H₁₃N₃O
Molecular Weight: 155,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20233070
CAS-Nummer: 1325164-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR