1-Ethyl-5-methyl-1H-pyrazole-3-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR306201-1G
APOSOR306201-1GEA 60.9 EUR
APOSOR306201-1G
1-Ethyl-5-methyl-1H-pyrazole-3-carboxylic acid 95+%
1-Ethyl-5-methyl-1H-pyrazole-3-carboxylic acid
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154.17 g/mol
Schmelzpunkt: 134…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08445953
CAS-Nummer: 50920-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR