1-(Pyridin-3-yl)-1H-pyrazole-4-carboxylic acid 98%

Lieferant: Apollo Scientific

OR306223-1G
APOSOR306223-1GEA 59.6 EUR
APOSOR306223-1G
1-(Pyridin-3-yl)-1H-pyrazole-4-carboxylic acid 98%
1-(Pyridin-3-yl)-1H-pyrazole-4-carboxylic acid
Formel: C₉H₇N₃O₂
Molecular Weight: 189.171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18260889
CAS-Nummer: 1014631-89-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR